This medium-duty, nuclear-grade duct tape features a polyethylene-coated cloth backing and an aggressive natural rubber adhesive designed for demanding applications in harsh environments, such as nuclear power plants, shipyards, and steel mills. Engineered to resist corrosion and provide superior holding power, it is suitable for bundling, hanging, sealing, and seaming tasks. The tape offers a balance of tensile strength and tearability, conforms well to irregular surfaces, and is designed to adhere to a wide variety of surfaces including rough materials.
- Adhesive type: Natural rubber
- Adhesion to steel: 80 oz./in.
- Tensile strength: 26 lb/in.
- Thickness: 11 mil
- Elongation: 20%
- Operating temperature range: 40°F to 200°F
- Complies with ASME NQA-1 and NRC Reg 1.38 standards
- Features easy unwind, curl resistance, and straight tear properties