Learn More
Grainger Nashua 357 Duct and Repair Tape, 13 mil, PE-Coated Cloth, 60 yd
Catalog No. 502753808
Encompass_Preferred
Nashua 357 is a premium 13 mil heavy-duty duct tape designed for demanding industrial applications. This single-coated, PE-coated cloth tape features a high-tack natural rubber adhesive that provides a secure bond to a variety of smooth and rough surfaces. It is engineered for permanent applications requiring high tensile strength and resistance to abrasion, punctures, and many common chemicals.
  • High tensile strength cloth backing for difficult bundling and repair tasks.
  • Waterproof construction resists moisture, solvents, and chemicals.
  • Abrasion and puncture resistant for added durability in tough environments.
  • Designed for a crisp, straight tear for clean application.
  • Wide operating temperature range of 4°C to 93°C (40°F to 200°F).
  • UL 723 and UL Listed for safety and performance compliance.
Catalog No. 50-275-3808 Supplier Grainger Supplier No. 15R447
Nashua 357 is a premium 13 mil heavy-duty duct tape designed for demanding industrial applications. This single-coated, PE-coated cloth tape features a high-tack natural rubber adhesive that provides a secure bond to a variety of smooth and rough surfaces. It is engineered for permanent applications requiring high tensile strength and resistance to abrasion, punctures, and many common chemicals.
  • High tensile strength cloth backing for difficult bundling and repair tasks.
  • Waterproof construction resists moisture, solvents, and chemicals.
  • Abrasion and puncture resistant for added durability in tough environments.
  • Designed for a crisp, straight tear for clean application.
  • Wide operating temperature range of 4°C to 93°C (40°F to 200°F).
  • UL 723 and UL Listed for safety and performance compliance.