Learn More
Grainger Nashua Duct Tape, Medium Duty, White, 1-7/8 in x 60 yd
Catalog No. 502756182
Encompass_Preferred
This medium duty duct tape is constructed with a polyethylene-coated cloth backing and a rubber-based pressure-sensitive adhesive, designed for versatile industrial and maintenance applications. It is engineered to conform well to irregular surfaces while providing reliable adhesion to a wide variety of materials. The tape is suitable for both indoor and outdoor use, featuring moisture resistance and an easy-unwind design to facilitate efficient application during duct sealing, bundling, and general repair tasks.
  • Designed for HVAC industry duct and equipment applications
  • Features an aggressive, permanent rubber adhesive
  • Compatible with a temperature range of 40°F to 200°F
  • Provides excellent tensile strength and tearability
  • Low VOC content and meets UL 723 standards
  • Moisture-resistant for reliable performance in various environments
Catalog No. 50-275-6182 Supplier Grainger Supplier No. 15R455
This medium duty duct tape is constructed with a polyethylene-coated cloth backing and a rubber-based pressure-sensitive adhesive, designed for versatile industrial and maintenance applications. It is engineered to conform well to irregular surfaces while providing reliable adhesion to a wide variety of materials. The tape is suitable for both indoor and outdoor use, featuring moisture resistance and an easy-unwind design to facilitate efficient application during duct sealing, bundling, and general repair tasks.
  • Designed for HVAC industry duct and equipment applications
  • Features an aggressive, permanent rubber adhesive
  • Compatible with a temperature range of 40°F to 200°F
  • Provides excellent tensile strength and tearability
  • Low VOC content and meets UL 723 standards
  • Moisture-resistant for reliable performance in various environments