Learn More
Grainger Nashua 357 Premium Duct Tape, PE-Coated Cloth, 13 Mil, 2 3/4 in. x 60 yd., White
Catalog No. 502757076
Encompass_Preferred
This premium 13 mil duct tape features a polyethylene-coated cloth backing and a high-tack rubber adhesive, designed for demanding HVAC, industrial, and maintenance applications. It provides excellent moisture, chemical, and abrasion resistance while maintaining a flexible, easy-to-handle profile that conforms well to irregular surfaces. This tape is suitable for both indoor and outdoor use in professional environments.
  • High tensile strength polyethylene-coated cloth backing for heavy-duty performance.
  • Rubber adhesive that offers high tack and flows over irregular surfaces.
  • Wide operating temperature range of 4°C to 93°C (40°F to 200°F).
  • Conforms to a wide variety of surfaces for reliable sealing and repair.
  • Hand tearable design with clean, straight tearing properties.
  • Low VOC content, contributing toward LEED EQ Credit 4.1.
Catalog No. 50-275-7076 Supplier Grainger Supplier No. 15R448
This premium 13 mil duct tape features a polyethylene-coated cloth backing and a high-tack rubber adhesive, designed for demanding HVAC, industrial, and maintenance applications. It provides excellent moisture, chemical, and abrasion resistance while maintaining a flexible, easy-to-handle profile that conforms well to irregular surfaces. This tape is suitable for both indoor and outdoor use in professional environments.
  • High tensile strength polyethylene-coated cloth backing for heavy-duty performance.
  • Rubber adhesive that offers high tack and flows over irregular surfaces.
  • Wide operating temperature range of 4°C to 93°C (40°F to 200°F).
  • Conforms to a wide variety of surfaces for reliable sealing and repair.
  • Hand tearable design with clean, straight tearing properties.
  • Low VOC content, contributing toward LEED EQ Credit 4.1.