Learn More
Grainger Nashua Duct Tape, PE-Coated Cloth, 11 Mil, White, 16/Case
Catalog No. 502757121
Encompass_Preferred
This 11 mil professional grade duct tape is designed for high-quality, premium maintenance applications across a variety of environments. Constructed with a PE-coated cloth carrier and a rubber-based adhesive, this medium-duty tape offers reliable performance for HVAC and general industrial use.
  • 11 mil thickness provides durability and strength
  • Rubber adhesive offers strong bonding
  • Low VOC content contributes to EQ credit 4.1 for low emitting materials
  • Operating temperature range of 40°F to 200°F
  • Color: White
  • Dimensions: 2 3/4 in. width by 60 yd length
Catalog No. 50-275-7121 Supplier Grainger Supplier No. 15R438
This 11 mil professional grade duct tape is designed for high-quality, premium maintenance applications across a variety of environments. Constructed with a PE-coated cloth carrier and a rubber-based adhesive, this medium-duty tape offers reliable performance for HVAC and general industrial use.
  • 11 mil thickness provides durability and strength
  • Rubber adhesive offers strong bonding
  • Low VOC content contributes to EQ credit 4.1 for low emitting materials
  • Operating temperature range of 40°F to 200°F
  • Color: White
  • Dimensions: 2 3/4 in. width by 60 yd length