Learn More
Grainger TapeCase General Purpose Duct Tape, Cotton Cloth, White, 2 in. x 60 yd., 1 ea.
Catalog No. 502754301
Encompass_Preferred
This continuous roll of white cloth tape is designed for general purpose applications including assembly, manufacturing, and maintenance. Featuring a rubber adhesive on an uncoated cotton cloth backing, this standard-duty tape provides reliable performance in both indoor and outdoor environments.
  • Electrically non-conductive
  • 10.5 mil thickness
  • 36 lb/in tensile strength
  • 32 oz/in adhesion to steel
  • Corrosion-resistant properties
  • Suitable for indoor and outdoor use
  • Continuous roll form
Catalog No. 50-275-4301 Supplier Grainger Supplier No. 15C777
This continuous roll of white cloth tape is designed for general purpose applications including assembly, manufacturing, and maintenance. Featuring a rubber adhesive on an uncoated cotton cloth backing, this standard-duty tape provides reliable performance in both indoor and outdoor environments.
  • Electrically non-conductive
  • 10.5 mil thickness
  • 36 lb/in tensile strength
  • 32 oz/in adhesion to steel
  • Corrosion-resistant properties
  • Suitable for indoor and outdoor use
  • Continuous roll form