Learn More
Grainger Polyken Cargo-Pit Seam Tape, Fiberglass Cloth, White, 3 in. x 36 yd., 12/Pk.
Catalog No. 502756385
Encompass_Preferred
This heavy-duty, flame-retardant fiberglass cloth tape is engineered for specialized aerospace applications, specifically for use in cargo pits on freight-carrying aircraft. It features a bromide-free acrylic adhesive that provides reliable performance across a broad temperature range while maintaining superior tensile strength.
  • Super high tensile strength of 134 lb/in.
  • Flame retardant design meeting FAR 25.853(a), 25.855(d), and 25.856(a) standards.
  • Excellent adhesion to steel at 45 oz./in.
  • Conformable backing allows for application on curves and corners.
  • Resists flagging and lifting for secure, long-term use.
  • Operational temperature range of -40°F to +275°F.
  • Compatible with Airbus (ABD0031) and Boeing (BMS 5-146) specifications.
Catalog No. 50-275-6385 Supplier Grainger Supplier No. 15R478
This heavy-duty, flame-retardant fiberglass cloth tape is engineered for specialized aerospace applications, specifically for use in cargo pits on freight-carrying aircraft. It features a bromide-free acrylic adhesive that provides reliable performance across a broad temperature range while maintaining superior tensile strength.
  • Super high tensile strength of 134 lb/in.
  • Flame retardant design meeting FAR 25.853(a), 25.855(d), and 25.856(a) standards.
  • Excellent adhesion to steel at 45 oz./in.
  • Conformable backing allows for application on curves and corners.
  • Resists flagging and lifting for secure, long-term use.
  • Operational temperature range of -40°F to +275°F.
  • Compatible with Airbus (ABD0031) and Boeing (BMS 5-146) specifications.