Learn More
Grainger Nashua Nuclear Grade Duct Tape, Yellow, Cloth, 11 mil, 60 yd
Catalog No. 502756105
Encompass_Preferred
This nuclear-grade duct tape features a polyethylene-coated cloth backing and a high-performance natural rubber adhesive, designed for sealing and repair applications in sensitive environments such as nuclear power plants, shipyards, and manufacturing facilities. The specialized adhesive system ensures effective bonding while remaining gentle on protective gear and surfaces. The tape offers excellent resistance to moisture, providing reliable holding power for tasks ranging from bundling and color-coding to securing critical safety materials.
  • Adhesion to steel of 80 oz./in.
  • Operating temperature range of 40°F to 200°F
  • Tensile strength of 26 lb./in.
  • Conforms well to irregular surfaces with 20% elongation
  • Meets ASME NQA-1 and NRC Reg 1.38 standards
  • Features low VOC content and contributes to LEED credit 4.1
  • Moisture-resistant, single-sided design
Catalog No. 50-275-6105 Supplier Grainger Supplier No. 15R445
This nuclear-grade duct tape features a polyethylene-coated cloth backing and a high-performance natural rubber adhesive, designed for sealing and repair applications in sensitive environments such as nuclear power plants, shipyards, and manufacturing facilities. The specialized adhesive system ensures effective bonding while remaining gentle on protective gear and surfaces. The tape offers excellent resistance to moisture, providing reliable holding power for tasks ranging from bundling and color-coding to securing critical safety materials.
  • Adhesion to steel of 80 oz./in.
  • Operating temperature range of 40°F to 200°F
  • Tensile strength of 26 lb./in.
  • Conforms well to irregular surfaces with 20% elongation
  • Meets ASME NQA-1 and NRC Reg 1.38 standards
  • Features low VOC content and contributes to LEED credit 4.1
  • Moisture-resistant, single-sided design