Learn More
Grainger Nashua Duct Tape, Yellow, 11 Mil, 1 7/8 in. x 60 yd.
Catalog No. 502754241
Encompass_Preferred
This professional grade, 11 mil thick duct tape features a polyethylene-coated cloth backing and natural rubber adhesive, providing an aggressive grip on rough surfaces and superior holding power for diverse applications. Designed for reliability in indoor and outdoor environments, this moisture- and curl-resistant tape is UL 723 listed and supports LEED® credits as a low VOC content product.
  • Features an excellent balance of tensile strength and tearability
  • Provides aggressive adhesion to rough surfaces
  • Suitable for bundling, hanging, holding, labeling, sealing, and seaming
  • Operates in a temperature range of 40°F to 200°F
  • Contributes toward satisfying EQ Credit 4.1 for low emitting materials
  • Available in a yellow, continuous roll format
Catalog No. 50-275-4241 Supplier Grainger Supplier No. 15R459
This professional grade, 11 mil thick duct tape features a polyethylene-coated cloth backing and natural rubber adhesive, providing an aggressive grip on rough surfaces and superior holding power for diverse applications. Designed for reliability in indoor and outdoor environments, this moisture- and curl-resistant tape is UL 723 listed and supports LEED® credits as a low VOC content product.
  • Features an excellent balance of tensile strength and tearability
  • Provides aggressive adhesion to rough surfaces
  • Suitable for bundling, hanging, holding, labeling, sealing, and seaming
  • Operates in a temperature range of 40°F to 200°F
  • Contributes toward satisfying EQ Credit 4.1 for low emitting materials
  • Available in a yellow, continuous roll format