Learn More
Grainger L.B. White Portable Gas Ducted Heater, 80,000 BtuH, Propane, 1 ea
Catalog No. 502765613
Encompass_Preferred
This portable, ductable direct gas-fired forced-air heater provides 80,000 BtuH of heating capacity for temporary heating in buildings under construction or repair, and is approved for USA tent heating applications with temporary human occupancy. Designed with a durable steel housing and Tri-shield finish, this heater delivers reliable performance via electronic spark ignition.
  • 99.97% fuel efficiency
  • 450 CFM air output for effective heat distribution
  • Remote thermostat with 1.4 ft. cord included
  • Integrated centrifugal blower for quiet operation
  • Includes gas hose and regulator
  • Dual high-limit switches and enclosed flame for safety
  • Vent-only mode available for air circulation
  • Lightweight, portable design with 27 hr max run time
Catalog No. 50-276-5613 Supplier Grainger Supplier No. 16W283
This portable, ductable direct gas-fired forced-air heater provides 80,000 BtuH of heating capacity for temporary heating in buildings under construction or repair, and is approved for USA tent heating applications with temporary human occupancy. Designed with a durable steel housing and Tri-shield finish, this heater delivers reliable performance via electronic spark ignition.
  • 99.97% fuel efficiency
  • 450 CFM air output for effective heat distribution
  • Remote thermostat with 1.4 ft. cord included
  • Integrated centrifugal blower for quiet operation
  • Includes gas hose and regulator
  • Dual high-limit switches and enclosed flame for safety
  • Vent-only mode available for air circulation
  • Lightweight, portable design with 27 hr max run time