Learn More
Grainger L.B. White Portable Gas Ducted Tent Heater, 350,000 BtuH, Dual Fuel, 1 ea
Catalog No. 502766347
Encompass_Preferred
This direct-fired, portable gas-ducted heater is designed for tent heating and remediation projects, providing a primary heat source in ventilated environments. The system features a dual-fuel design capable of operating on either LP or natural gas and includes a centrifugal blower for quiet performance. It is certified for use in U.S. and Canadian tent heating applications requiring temporary human occupancy.
  • Heating capacity output of 350,000 BtuH
  • Dual fuel model allows operation on LP or NG
  • Features a centrifugal blower for very quiet operation
  • Includes gas hose, regulator, and remote thermostat
  • Equipped with two high-limit switches for enhanced safety
  • Fan-only operation mode available
  • Self-diagnostic ignition control board
  • 99.97% fuel efficiency
Catalog No. 50-276-6347 Supplier Grainger Supplier No. 16W290
Add to Cart
Add to Cart
Request a Quote
This direct-fired, portable gas-ducted heater is designed for tent heating and remediation projects, providing a primary heat source in ventilated environments. The system features a dual-fuel design capable of operating on either LP or natural gas and includes a centrifugal blower for quiet performance. It is certified for use in U.S. and Canadian tent heating applications requiring temporary human occupancy.
  • Heating capacity output of 350,000 BtuH
  • Dual fuel model allows operation on LP or NG
  • Features a centrifugal blower for very quiet operation
  • Includes gas hose, regulator, and remote thermostat
  • Equipped with two high-limit switches for enhanced safety
  • Fan-only operation mode available
  • Self-diagnostic ignition control board
  • 99.97% fuel efficiency