Learn More
Grainger Ducting with Adapter Steel 18 dia
Catalog No. 502766293
Encompass_Preferred
The L.B. White Portable Gas Heater Duct Kit is designed for use with L.B. White portable gas heaters to provide heated air through ductwork. It includes a blower assembly and mounting hardware to attach it to the heater. The blower provides 585 CFM airflow and can heat areas up to 2000 square feet when paired with a compatible heater. This duct kit allows you to distribute heat to multiple rooms or areas from a single portable propane or natural gas heater. It's made of heavy-gauge steel and is ETL approved.
Catalog No. 50-276-6293 Supplier Grainger Supplier No. 16W311
The L.B. White Portable Gas Heater Duct Kit is designed for use with L.B. White portable gas heaters to provide heated air through ductwork. It includes a blower assembly and mounting hardware to attach it to the heater. The blower provides 585 CFM airflow and can heat areas up to 2000 square feet when paired with a compatible heater. This duct kit allows you to distribute heat to multiple rooms or areas from a single portable propane or natural gas heater. It's made of heavy-gauge steel and is ETL approved.