Learn More
Caron Products And Services Ductless 1000mm Fume Cabinet, 3 Layer Chemcap Filtration
Catalog No. NC3690090
Encompass_Preferred
Ductless 1000mm Fume Cabinet, 3 Layer Chemcap Filtration
Catalog No. NC3690090 Supplier Caron Products And Services Supplier No. PR10044
Add to Cart
Add to Cart
Request a Quote
Ductless 1000mm Fume Cabinet, 3 Layer Chemcap Filtration