Learn More
Air Science Usa Ductless Fume Hood 39" Wide
Supplier: Air Science Usa P15XTA
The Advanced Series ductless fume hoods are a series of high efficiency products designed to protect the user and the environment from hazardous vapors generated on the work surface. At the heart of the Purair ductless fume hood product line is the innovative Multiplex Filtration Technology that creates a safe work environment over the widest range of applications in the industry. Unit offers high operator protection to fume and particle hazards. Energy efficient fan delivers face velocity of 100 fpm. A blocked filter alarm warns of insufficient face velocity. Unit requires one filter sold separately: Astm-001 filter for solvents, Astm-015 for acids, Astm-012 for ammonia, Astm-013 for formaldehydes, or Astm-030 HEPA filter for particulates. Optional Secondary Asts-001, Asts-015, Asts-013, Asts-030 back-up filters can also be added for improved safety