Learn More
Air Science Usa Ductless Fume Hood 69" Wide Advanced Triple Blower
SDP
Catalog No. NC3948030
Encompass_Preferred
PurairADVANCED Ductless Fume Hood, Triple Blower Version, 69" / 1800mm Nominal Width, Comprising of a P40-HEAD-A, P30-XT-ENCL-TRIP, TRAY-P30, and DWYER Airflow Meter, Standard Depth, 115V 60Hz, North American Cord set (unless specified). Requires Minimum Three ASTM-XXX Main Filters and Three Optional ASTS-XXX Safety Filters
Catalog No. NC3948030 Supplier Air Science Usa Supplier No. P30XTTRIPAADVP
PurairADVANCED Ductless Fume Hood, Triple Blower Version, 69" / 1800mm Nominal Width, Comprising of a P40-HEAD-A, P30-XT-ENCL-TRIP, TRAY-P30, and DWYER Airflow Meter, Standard Depth, 115V 60Hz, North American Cord set (unless specified). Requires Minimum Three ASTM-XXX Main Filters and Three Optional ASTS-XXX Safety Filters