Learn More
Research Products International Corp DuelingSyringes Tissue Disperser with Screen, 5 Sets
SDP
Catalog No. 501537760
Encompass
Unique DuelingSyringes are designed to disperse tissues using a low shear method to recover intact, viable cells for soft tissues (brain, liver, muscle). Ideal for isolation of high quality intacellular organelles. DuelingSyringes offer a faster and gentler method of processing small soft tissue samples.
Tissue Dispenser features a pair of 1 ml syringes that are connected together via their Luer connections. One syringe is embedded with a 500µm stainless steel screen of low shear samples dispersion. Easy to use - Load tissue into sample syringe (embedded with screen), add 0.3-1ml dispersion media (buffer, growth media) into receiving syringe, reconnect syringes and push sample back and forth from syringe to syringe.
Catalog No. 50-153-7760 Supplier Research Products International Corp Supplier No. 299116
Unique DuelingSyringes are designed to disperse tissues using a low shear method to recover intact, viable cells for soft tissues (brain, liver, muscle). Ideal for isolation of high quality intacellular organelles. DuelingSyringes offer a faster and gentler method of processing small soft tissue samples.
Tissue Dispenser features a pair of 1 ml syringes that are connected together via their Luer connections. One syringe is embedded with a 500µm stainless steel screen of low shear samples dispersion. Easy to use - Load tissue into sample syringe (embedded with screen), add 0.3-1ml dispersion media (buffer, growth media) into receiving syringe, reconnect syringes and push sample back and forth from syringe to syringe.