Learn More
Selleck Chemical LLC Dulaglutide-5mg
SDP
Supplier:  Selleck Chemical LLC P11155MG
Encompass_Preferred
Dulaglutide is a GLP-1 receptor agonist approved for the management of hyper glycaemia in people with type 2 diabetes in many countries. P1115 is its biologically active polypeptide part, the sequence is His-Gly-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser- Tyr-Leu-Glu-Glu-Gln-Ala-Ala-Lys-Glu-Phe-Ile-Ala-Trp-Leu-Val-Lys-Gly-Gly-Gly.
Catalog No. 50-238-9673