Learn More
Sigma Aldrich Fine Chemicals Biosciences Dulbecco's Modified Eagle's Medium - high glucose With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture | 500ML
Catalog No. NC1908374
Encompass
Dulbecco's Modified Eagle's Medium - high glucose With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture | 500ML
Catalog No. NC1908374 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D6546500ML
Dulbecco's Modified Eagle's Medium - high glucose With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture | 500ML