Learn More
Promocell GmbH PromoCell Dulbeccos PBS 500 ml, w/o Ca++/ Mg++
Catalog No. 50306365
Encompass
Buffers and Salt Solutions are frequently used in tissue culture, cell isolation procedures, and molecular biology.For convenience, PromoCell offers ready-to-use Buffers and Salt Solutions. They are prepared from raw materials of the highest quality using tissue culture grade water.Dulbecco's PBS can be purchased with or without Ca^2 ^/ Mg^2 ^.
Catalog No. 50-306-365 Supplier Promocell GmbH Supplier No. C40232
Buffers and Salt Solutions are frequently used in tissue culture, cell isolation procedures, and molecular biology.For convenience, PromoCell offers ready-to-use Buffers and Salt Solutions. They are prepared from raw materials of the highest quality using tissue culture grade water.Dulbecco's PBS can be purchased with or without Ca^2 ^/ Mg^2 ^.