Learn More
Promocell GmbH PromoCell Dulbeccos PBS pH 7,2-7,8 500 ml, Standard, with Ca++/ Mg++
Catalog No. 50306364
Encompass
Buffers and Salt Solutions are frequently used in tissue culture, cell isolation procedures, and molecular biology.For convenience, PromoCell offers ready-to-use Buffers and Salt Solutions. They are prepared from raw materials of the highest quality using tissue culture grade water.Dulbecco's PBS can be purchased with or without Ca^2 ^/ Mg^2 ^.
Catalog No. 50-306-364 Supplier Promocell GmbH Supplier No. C40230
Add to Cart
Add to Cart
Buffers and Salt Solutions are frequently used in tissue culture, cell isolation procedures, and molecular biology.For convenience, PromoCell offers ready-to-use Buffers and Salt Solutions. They are prepared from raw materials of the highest quality using tissue culture grade water.Dulbecco's PBS can be purchased with or without Ca^2 ^/ Mg^2 ^.