Learn More
Sigma Aldrich Fine Chemicals Biosciences DULBECCOS MOD EGLS MED,6X500ML
Catalog No. NC1607187
Encompass
Dulbecco’s Modified Eagle’s Medium - high glucose With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture6 x 500ml
Catalog No. NC1607187 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D57966X500ML
Dulbecco’s Modified Eagle’s Medium - high glucose With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture6 x 500ml