Learn More
HIMEDIA Dulbecco’s Modified Eagle Medium/ Nutrient Mixture F-12 Ham (DMEM/ F12, 3:1 Mixture), High glucose w/ L-Glutamine w/o Calcium chloride, HEPES buffer and Sodium bicarbonate
Catalog No. NC1624508
Encompass_Preferred
Dulbecco’s Modified Eagle Medium/ Nutrient Mixture F-12 Ham (DMEM/ F12, 3:1 Mixture), High glucose w/ L-Glutamine w/o Calcium chloride, HEPES buffer and Sodium bicarbonate
This item is not returnable. View return policy
Catalog No. NC1624508 Supplier HIMEDIA Supplier No. AT1895L
Dulbecco’s Modified Eagle Medium/ Nutrient Mixture F-12 Ham (DMEM/ F12, 3:1 Mixture), High glucose w/ L-Glutamine w/o Calcium chloride, HEPES buffer and Sodium bicarbonate