Learn More
Sigma Aldrich Fine Chemicals Biosciences Dulbeccos Modified Eagles
Catalog No. 502901890
Encompass
The most basic formulation offered. This formulation is used by investigators who want to start with the essential components of DME and have the flexibility to optimize the formula for their own application.
Catalog No. 50-290-1890 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D503010L
The most basic formulation offered. This formulation is used by investigators who want to start with the essential components of DME and have the flexibility to optimize the formula for their own application.