Learn More
Bioworld Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham w/L-glutamine & sodium bicarbonate, w/o, 500 mL
Supplier:  Bioworld 306110991
Encompass_Preferred
With L-glutamine and sodium bicarbonate, without HEPES, liquid, sterile-filtered, cell culture tested.SpecificationsSynonyms: DME/F-12, 1:1 mixtureSterility: sterile-filteredForm: liquid with phenol redTotal impurities: Endotoxin, tested.
Catalog No. 50-254-216