Learn More
Sigma Aldrich Fine Chemicals Biosciences Dulbeccos Modified Eagles Medium - high glucose With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Catalog No. 501657368
Encompass
Dulbeccos Modified Eagles Medium - high glucose With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Catalog No. 50-165-7368 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D57966X1L
Dulbeccos Modified Eagles Medium - high glucose With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture