Learn More
Sigma Aldrich Fine Chemicals Biosciences Dulbeccos Modified Eagles1L
Catalog No. 502901904
Encompass
Suitable for growth of a wide range of cell types in low serum conditions. The removal of phenol red is often preferred by investigators working with clonal and stem cell cultures.
Catalog No. 50-290-1904 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D290610X1L
Suitable for growth of a wide range of cell types in low serum conditions. The removal of phenol red is often preferred by investigators working with clonal and stem cell cultures.