Learn More
Sigma Aldrich Fine Chemicals Biosciences Dulbeccos Modified Eagles1L
Catalog No. 502905957
Encompass
This DMEM-Low glucose medium is a 1x complete medium with sodium pyruvate added. It also differs from the original DMEM-Hi formulation wherein pyridoxine is substituted for pyridoxal. Pyridoxal is an unstable component of media.
Catalog No. 50-290-5957 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D60466X1L
This DMEM-Low glucose medium is a 1x complete medium with sodium pyruvate added. It also differs from the original DMEM-Hi formulation wherein pyridoxine is substituted for pyridoxal. Pyridoxal is an unstable component of media.