Learn More
Sigma Aldrich Fine Chemicals Biosciences Dulbeccos Modified Eagles1L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D57961L
Encompass
This DMEM-Hi glucose medium is a 1x complete medium with no added factors (common modifications) such as L-alanyl-L-glutamine HEPES or sodium pyruvate. It differs from the original DMEM-Hi formulation wherein pyridoxine is substituted for pyridoxal. Pyridoxal is an unstable component of media.
Catalog No. 50-290-0045