Learn More
Sigma Aldrich Fine Chemicals Biosciences Dulbeccos Modified Eagles500ML
Catalog No. 502901493
Encompass
Pyridoxal which is an aldehyde has been replaced with pyridoxine. This modification has been shown to improve the stability of the medium. Suitable for growth of a wide range of cell types in low serum conditions. Absence of phenol red makes this a preferred medium for clonal and stem cell culture.
Catalog No. 50-290-1493 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D6434500ML
Pyridoxal which is an aldehyde has been replaced with pyridoxine. This modification has been shown to improve the stability of the medium. Suitable for growth of a wide range of cell types in low serum conditions. Absence of phenol red makes this a preferred medium for clonal and stem cell culture.