Learn More
Sigma Aldrich Fine Chemicals Biosciences Dulbeccos Modified Eagles500ML
Catalog No. 502905966
Encompass
A measured mixture of chemically defined ingredients makes up a basal medium which is a type of complex media. DMEM is a medium supplemented with serum and sometimes with tryptose phosphate broth (TPB). DMEM/f-12 is a completely defined synthetic medium which is protein-free.
Catalog No. 50-290-5966 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D64216X500ML
A measured mixture of chemically defined ingredients makes up a basal medium which is a type of complex media. DMEM is a medium supplemented with serum and sometimes with tryptose phosphate broth (TPB). DMEM/f-12 is a completely defined synthetic medium which is protein-free.