Learn More
Sigma Aldrich Fine Chemicals Biosciences Dulbeccos Modified Eagles500ML
Catalog No. 502901913
Encompass
This DMEM-Hi glucose medium is a 1x complete medium with no added factors (common modifications) such as L-alanyl-L-glutamine HEPES or sodium pyruvate. It differs from the original DMEM-Hi formulation wherein pyridoxine is substituted for pyridoxal. Pyridoxal is an unstable component of media. This medium requires supplementation with L-glutamine or L-alanyl-L-glutamine.
Catalog No. 50-290-1913 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D56716X500ML
This DMEM-Hi glucose medium is a 1x complete medium with no added factors (common modifications) such as L-alanyl-L-glutamine HEPES or sodium pyruvate. It differs from the original DMEM-Hi formulation wherein pyridoxine is substituted for pyridoxal. Pyridoxal is an unstable component of media. This medium requires supplementation with L-glutamine or L-alanyl-L-glutamine.