Learn More
Sigma Aldrich Fine Chemicals Biosciences Dulbeccos Modified Eagles500ML
Catalog No. 502905193
Encompass
This DMEM-Hi glucose medium is a 1x complete medium with sodium pyruvate added. It also differs from the original DMEM-Hi formulation wherein pyridoxine is substituted for pyridoxal. Pyridoxal is an unstable component of media. This medium requires supplementation with L-glutamine or L-alanyl-L-glutamine.
Catalog No. 50-290-5193 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D65466X500ML
This DMEM-Hi glucose medium is a 1x complete medium with sodium pyruvate added. It also differs from the original DMEM-Hi formulation wherein pyridoxine is substituted for pyridoxal. Pyridoxal is an unstable component of media. This medium requires supplementation with L-glutamine or L-alanyl-L-glutamine.