Learn More
Sigma Aldrich Fine Chemicals Biosciences Dulbeccos Modified Eagles500ML
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D64296X500ML
Encompass
This DMEM-Hi glucose medium is a 1x complete medium with sodium pyruvate added. It also differs from the original DMEM-Hi formulation wherein pyridoxine is substituted for pyridoxal. Pyridoxal is an unstable component of media.
Catalog No. 50-290-0943