Learn More
ATCC Dulbecco's PBS w/o Ca or Mg; 500 ML
Catalog No. 502384473
Encompass
Dulbecco's Phosphate Buffered Saline (D-PBS) is used for washing cells, for use as a diluent, or for use as an inorganic base for standard media preparation. Formulation contains no calcium or magnesium. It has applications for cell culture, cell growth, and viability.
Catalog No. 50-238-4473 Supplier ATCC Supplier No. 302200
Dulbecco's Phosphate Buffered Saline (D-PBS) is used for washing cells, for use as a diluent, or for use as an inorganic base for standard media preparation. Formulation contains no calcium or magnesium. It has applications for cell culture, cell growth, and viability.