Learn More
Medchemexpress LLC HY-Y0418 100mg Medchemexpress, Dulcite CAS:608-66-2 Purity:>98%
SDP
Catalog No. 501970762
Encompass_Preferred
Medchemexpress, HY-Y0418 100mg Dulcite CAS:608-66-2 Dulcite is a sugar alcohol with a slightly sweet taste which is a metabolic breakdown product of galactose. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0762 Supplier Medchemexpress LLC Supplier No. HYY0418100MG
Medchemexpress, HY-Y0418 100mg Dulcite CAS:608-66-2 Dulcite is a sugar alcohol with a slightly sweet taste which is a metabolic breakdown product of galactose. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.