Learn More
Aviva Systems Biology Corp DULLARD Antibody - middle region (ARP47144_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47144_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DULLARD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DULLARD. Alternate names: HSA011916, NET56, DULLARDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59472