Learn More
Medchemexpress LLC Duloxetine | 116539-59-4 | 99.9% | 25 MG
SDP
Catalog No. 5000405808
Encompass_Preferred
Duloxetine, also known as LY248686, is a potent serotonin-norepinephrine reuptake inhibitor with a Ki of 4.6 nM. It is primarily utilized in research for its role in treating major depressive disorder and generalized anxiety disorder (GAD). This compound selectively inhibits the reuptake of serotonin and norepinephrine within the central nervous system, demonstrating less potency in inhibiting dopamine reuptake. It is intended for research use only and not for patient administration.
  • Serotonin-norepinephrine reuptake inhibitor
  • Used for major depressive disorder research
  • Used for generalized anxiety disorder research
  • Selective reuptake inhibitor at 5-HT and NA transporters
  • No significant affinity for various other receptors (dopaminergic, adrenergic, cholinergic, histaminergic, opioid, glutamate, GABA)
Catalog No. 50-004-05808 Supplier Medchemexpress LLC Supplier No. 25MGHYB0161
Duloxetine, also known as LY248686, is a potent serotonin-norepinephrine reuptake inhibitor with a Ki of 4.6 nM. It is primarily utilized in research for its role in treating major depressive disorder and generalized anxiety disorder (GAD). This compound selectively inhibits the reuptake of serotonin and norepinephrine within the central nervous system, demonstrating less potency in inhibiting dopamine reuptake. It is intended for research use only and not for patient administration.
  • Serotonin-norepinephrine reuptake inhibitor
  • Used for major depressive disorder research
  • Used for generalized anxiety disorder research
  • Selective reuptake inhibitor at 5-HT and NA transporters
  • No significant affinity for various other receptors (dopaminergic, adrenergic, cholinergic, histaminergic, opioid, glutamate, GABA)