Learn More
Medchemexpress LLC Duloxetine | 116539-59-4 | 99.9% | 5 MG
SDP
Catalog No. 5000406253
Encompass_Preferred
Duloxetine is a serotonin-norepinephrine reuptake inhibitor with a Ki of 4.6 nM. It is used for the treatment of major depressive disorder and generalized anxiety disorder (GAD). It inhibits the reuptake of serotonin and norepinephrine in the central nervous system and is a less potent inhibitor of dopamine reuptake. Its analgesic properties are attributed to sodium ion channel blockade.
  • Selective reuptake inhibitor at 5-HT and NA transporters
  • Less potent inhibitor of dopamine reuptake
  • No significant affinity for dopaminergic, adrenergic, cholinergic, histaminergic, opioid, glutamate, and GABA receptors
  • Potential to decrease pro-inflammatory cytokine activity and increase anti-inflammatory cytokines
  • Analgesic properties for conditions like diabetic neuropathy and fibromyalgia
Catalog No. 50-004-06253 Supplier Medchemexpress LLC Supplier No. HYB01615MG
Duloxetine is a serotonin-norepinephrine reuptake inhibitor with a Ki of 4.6 nM. It is used for the treatment of major depressive disorder and generalized anxiety disorder (GAD). It inhibits the reuptake of serotonin and norepinephrine in the central nervous system and is a less potent inhibitor of dopamine reuptake. Its analgesic properties are attributed to sodium ion channel blockade.
  • Selective reuptake inhibitor at 5-HT and NA transporters
  • Less potent inhibitor of dopamine reuptake
  • No significant affinity for dopaminergic, adrenergic, cholinergic, histaminergic, opioid, glutamate, and GABA receptors
  • Potential to decrease pro-inflammatory cytokine activity and increase anti-inflammatory cytokines
  • Analgesic properties for conditions like diabetic neuropathy and fibromyalgia