Learn More
Medchemexpress LLC Duloxetine | 116539-59-4 | 99.9% | 50 MG
SDP
Catalog No. 5000406078
Encompass_Preferred
Duloxetine is a serotonin-norepinephrine reuptake inhibitor with a Ki of 4.6 nM. It is used for the treatment of major depressive disorder and generalized anxiety disorder (GAD). For research use only; not sold to patients. Duloxetine inhibits the reuptake of serotonin and norepinephrine in the central nervous system. It is also considered a less potent inhibitor of dopamine reuptake. Duloxetine has no significant affinity for dopaminergic, adrenergic, cholinergic, histaminergic, opioid, glutamate, and GABA receptors, making it a selective reuptake inhibitor at the 5-HT and NA transporters.
  • Inhibits serotonin and norepinephrine reuptake
  • Treats major depressive disorder and generalized anxiety disorder
  • Acts as a less potent inhibitor of dopamine reuptake
  • Functions as a selective reuptake inhibitor at 5-HT and NA transporters
  • For research use only
Catalog No. 50-004-06078 Supplier Medchemexpress LLC Supplier No. HYB016150MG
Duloxetine is a serotonin-norepinephrine reuptake inhibitor with a Ki of 4.6 nM. It is used for the treatment of major depressive disorder and generalized anxiety disorder (GAD). For research use only; not sold to patients. Duloxetine inhibits the reuptake of serotonin and norepinephrine in the central nervous system. It is also considered a less potent inhibitor of dopamine reuptake. Duloxetine has no significant affinity for dopaminergic, adrenergic, cholinergic, histaminergic, opioid, glutamate, and GABA receptors, making it a selective reuptake inhibitor at the 5-HT and NA transporters.
  • Inhibits serotonin and norepinephrine reuptake
  • Treats major depressive disorder and generalized anxiety disorder
  • Acts as a less potent inhibitor of dopamine reuptake
  • Functions as a selective reuptake inhibitor at 5-HT and NA transporters
  • For research use only