Learn More
Medchemexpress LLC Duloxetine | 116539-59-4 | MFCD06801358 | 99.9% | 297.4 g/mol | C18H19NOS | 10 MG
SDP
Catalog No. 5000298836
Encompass_Preferred
Duloxetine is a serotonin-norepinephrine reuptake inhibitor (SNRI) used in research on major depressive disorder, generalized anxiety disorder, and related indications. It selectively inhibits serotonin and norepinephrine transporters (reported Ki ≈ 4.6 nM) and shows minimal affinity for many other receptor classes. Supplied as a research-grade compound with reported purity of 99.92%.
  • Selective serotonin-norepinephrine reuptake inhibitor with Ki ≈ 4.6 nM.
  • High reported purity (99.9%).
  • Available in small-mass packs and solution formats for research use.
  • Suitable for in vitro pharmacology and biochemical assays.
  • CAS number 116539-59-4 for unambiguous identification.
Catalog No. 50-002-98836 Supplier Medchemexpress LLC Supplier No. HYB016110MG
Duloxetine is a serotonin-norepinephrine reuptake inhibitor (SNRI) used in research on major depressive disorder, generalized anxiety disorder, and related indications. It selectively inhibits serotonin and norepinephrine transporters (reported Ki ≈ 4.6 nM) and shows minimal affinity for many other receptor classes. Supplied as a research-grade compound with reported purity of 99.92%.
  • Selective serotonin-norepinephrine reuptake inhibitor with Ki ≈ 4.6 nM.
  • High reported purity (99.9%).
  • Available in small-mass packs and solution formats for research use.
  • Suitable for in vitro pharmacology and biochemical assays.
  • CAS number 116539-59-4 for unambiguous identification.