Learn More
Selleck Chemical LLC Duloxetine S5071-1g
SDP
Catalog No. 503112132
Encompass_Preferred
Duloxetine (LY-248686) is a potent inhibitor of serotonin (5-hydroxytryptamine 5-HT) and noradrenaline (NE) uptake in vitro and in vivo and is 3- to 5-times more effective at inhibiting 5-HT uptake
Catalog No. 50-311-2132 Supplier Selleck Chemical LLC Supplier No. S50711G
Duloxetine (LY-248686) is a potent inhibitor of serotonin (5-hydroxytryptamine 5-HT) and noradrenaline (NE) uptake in vitro and in vivo and is 3- to 5-times more effective at inhibiting 5-HT uptake