Learn More
Medchemexpress LLC Duloxetine-d7 | 919514-01-5 | 97.3% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0161S1MG
Encompass_Preferred
Duloxetine-d7 is the deuterium labeled form of Duloxetine, a serotonin-norepinephrine reuptake inhibitor with a Ki of 4.6 nM. It is used for the treatment of major depressive disorder and generalized anxiety disorder (GAD). Stable heavy isotopes, such as deuterium, are incorporated into drug molecules for use as tracers in drug development and to potentially affect pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-07238