Learn More
Medchemexpress LLC HY-B0161A 100mg , Duloxetine (hydrochloride) CAS:136434-34-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0161A100MG
Encompass_Preferred
HY-B0161A 100mg Duloxetine (hydrochloride) CAS: 136434-34-9 Duloxetine hydrochloride ((S)-Duloxetine hydrochloride) is a serotonin-norepinephrine reuptake inhibitor (SNRI) with a Ki of 4.6 nM, used for treatment of major depressive disorder and generalized anxiety disorder (GAD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1119