Learn More
Selleck Chemical LLC Duloxetine HCl S2084-100mg
SDP
Catalog No. 503110651
Encompass_Preferred
Duloxetine HCl (LY-248686) is a serotonin-norepinephrine reuptake inhibitor with Ki of 4 6 nM used for treatment of major depressive disorder and generalized anxiety disorder (GAD)
Catalog No. 50-311-0651 Supplier Selleck Chemical LLC Supplier No. S2084100MG
Duloxetine HCl (LY-248686) is a serotonin-norepinephrine reuptake inhibitor with Ki of 4 6 nM used for treatment of major depressive disorder and generalized anxiety disorder (GAD)