Learn More
Selleck Chemical LLC Duloxetine HCl S2084-10mg
SDP
Catalog No. 503111264
Encompass_Preferred
Duloxetine HCl (LY-248686) is a serotonin-norepinephrine reuptake inhibitor with Ki of 4 6 nM used for treatment of major depressive disorder and generalized anxiety disorder (GAD)
Catalog No. 50-311-1264 Supplier Selleck Chemical LLC Supplier No. S208410MG
Duloxetine HCl (LY-248686) is a serotonin-norepinephrine reuptake inhibitor with Ki of 4 6 nM used for treatment of major depressive disorder and generalized anxiety disorder (GAD)