Learn More
TARGETMOL CHEMICALS INC Duloxetine hydrochloride 100MG
SDP
Catalog No. 502878586
Encompass_Preferred
Also available in 1 mL, 10 mg, 50 mg, 200 mg and bulk. Please contact Fisher for quotes. Duloxetine hydrochloride ((S)-Duloxetine hydrochloride) is a thiophene derivative and selective neurotransmitter uptake inhibitor for serotonin and noradrenaline (SNRI). It is an antidepressant agent and anxiolytic, also used for treating pain in patients with diabetes mellitus and fibromyalgia. Purity 99.97%
Catalog No. 50-287-8586 Supplier TARGETMOL CHEMICALS INC Supplier No. T1471100MG
Also available in 1 mL, 10 mg, 50 mg, 200 mg and bulk. Please contact Fisher for quotes. Duloxetine hydrochloride ((S)-Duloxetine hydrochloride) is a thiophene derivative and selective neurotransmitter uptake inhibitor for serotonin and noradrenaline (SNRI). It is an antidepressant agent and anxiolytic, also used for treating pain in patients with diabetes mellitus and fibromyalgia. Purity 99.97%