Learn More
Medchemexpress LLC Duloxetine (hydrochloride) | 136434-34-9 | >99.9% | 5 MG
SDP
Catalog No. 5000406907
Encompass_Preferred
Duloxetine hydrochloride is a chemical compound that functions as a serotonin-norepinephrine reuptake inhibitor (SNRI). It is primarily used in research contexts for studying major depressive disorder and generalized anxiety disorder (GAD).
  • Functions as a serotonin-norepinephrine reuptake inhibitor (SNRI)
  • Has a Ki of 4.6 nM
  • Used for treatment of major depressive disorder
  • Used for generalized anxiety disorder (GAD)
  • For research use only
Catalog No. 50-004-06907 Supplier Medchemexpress LLC Supplier No. HYB0161A5MG
Duloxetine hydrochloride is a chemical compound that functions as a serotonin-norepinephrine reuptake inhibitor (SNRI). It is primarily used in research contexts for studying major depressive disorder and generalized anxiety disorder (GAD).
  • Functions as a serotonin-norepinephrine reuptake inhibitor (SNRI)
  • Has a Ki of 4.6 nM
  • Used for treatment of major depressive disorder
  • Used for generalized anxiety disorder (GAD)
  • For research use only