Learn More
Medchemexpress LLC Duloxetine hydrochloride | 136434-34-9 | 99.97% | 1 ML
SDP
Catalog No. 5000406935
Encompass_Preferred
Duloxetine hydrochloride ((S)-Duloxetine hydrochloride) is a serotonin-norepinephrine reuptake inhibitor (SNRI) used for the treatment of major depressive disorder and generalized anxiety disorder (GAD). This product is for research use only and not sold to patients.
  • Serotonin-norepinephrine reuptake inhibitor (SNRI)
  • Used for major depressive disorder and generalized anxiety disorder
  • For research use only
Catalog No. 50-004-06935 Supplier Medchemexpress LLC Supplier No. HYB0161A10MM/1ML
Duloxetine hydrochloride ((S)-Duloxetine hydrochloride) is a serotonin-norepinephrine reuptake inhibitor (SNRI) used for the treatment of major depressive disorder and generalized anxiety disorder (GAD). This product is for research use only and not sold to patients.
  • Serotonin-norepinephrine reuptake inhibitor (SNRI)
  • Used for major depressive disorder and generalized anxiety disorder
  • For research use only