Learn More
Medchemexpress LLC Duloxetine hydrochloride | 136434-34-9 | MFCD06407958 | >99.0% | 333.9 g/mol | C18H19NOS·HCl | 10 MG
SDP
Catalog No. 5000298837
Encompass_Preferred
Duloxetine hydrochloride is the hydrochloride salt of duloxetine, a serotonin-norepinephrine reuptake inhibitor (SNRI) commonly used in research on mood disorders, anxiety, and neuropathic pain. The compound is supplied for laboratory research at research-grade purity and is used in biochemical and pharmacological assays.
  • Serotonin-norepinephrine reuptake inhibitor activity with reported Ki ≈ 4.6 nM.
  • Hydrochloride salt form for improved stability and handling.
  • High purity suitable for research (typically ≥99% reported by suppliers).
  • Used in studies of major depressive disorder, generalized anxiety disorder, and neuropathic pain mechanisms.
  • Small research pack sizes available for preclinical and analytical work.
Catalog No. 50-002-98837 Supplier Medchemexpress LLC Supplier No. HYB0161A10MG
Duloxetine hydrochloride is the hydrochloride salt of duloxetine, a serotonin-norepinephrine reuptake inhibitor (SNRI) commonly used in research on mood disorders, anxiety, and neuropathic pain. The compound is supplied for laboratory research at research-grade purity and is used in biochemical and pharmacological assays.
  • Serotonin-norepinephrine reuptake inhibitor activity with reported Ki ≈ 4.6 nM.
  • Hydrochloride salt form for improved stability and handling.
  • High purity suitable for research (typically ≥99% reported by suppliers).
  • Used in studies of major depressive disorder, generalized anxiety disorder, and neuropathic pain mechanisms.
  • Small research pack sizes available for preclinical and analytical work.