Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Duloxetine metabolite Para-Naphthol Duloxetine | 949095-98-1 | 297.41 | 50 MG
SDP

Catalog No. 5000365979
Encompass_Preferred

Duloxetine metabolite Para-Naphthol Duloxetine is a metabolite of Duloxetine, a serotonin-norepinephrine reuptake inhibitor (SNRI). This product is intended for research use only and is not sold to patients.

  • Metabolite of Duloxetine
  • For research use only
  • Molecular formula: C18H19NOS
  • Appearance: Solid
  • Color: White to off-white
  • Store powder at -20°C for 3 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month

Catalog No. 50-003-65979 Supplier Medchemexpress LLC Supplier No. 50MGHYG0005
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Duloxetine metabolite Para-Naphthol Duloxetine is a metabolite of Duloxetine, a serotonin-norepinephrine reuptake inhibitor (SNRI). This product is intended for research use only and is not sold to patients.

  • Metabolite of Duloxetine
  • For research use only
  • Molecular formula: C18H19NOS
  • Appearance: Solid
  • Color: White to off-white
  • Store powder at -20°C for 3 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.